Anti-CXCL10 Mouse Monoclonal Antibody [clone: 163426]

Supplier: R & D Systems
MAB1117
220018-809EA 772.25 USD
220018-809
Anti-CXCL10 Mouse Monoclonal Antibody [clone: 163426]
Antibodies
Type: Primary
Antigen: CXCL10
Clonality: Monoclonal
Clone: 163426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR