Anti-CXCL1/2/3 Mouse Monoclonal Antibody [clone: 186413]

Supplier: R & D Systems
MAB1114
220018-807EA 763.57 USD
220018-807
Anti-CXCL1/2/3 Mouse Monoclonal Antibody [clone: 186413]
Antibodies
Type: Primary
Antigen: CXCL1/2/3
Clonality: Monoclonal
Clone: 186413
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR