Anti-PRLR Mouse Monoclonal Antibody [clone: 218623]

Supplier: R & D Systems
MAB11121
220018-806EA 763.57 USD
220018-806
Anti-PRLR Mouse Monoclonal Antibody [clone: 218623]
Antibodies
Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: 218623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR