Anti-PRLR Mouse Monoclonal Antibody [clone: 218616]

Supplier: R & D Systems
MAB1112
220018-805EA 772.25 USD
220018-805
Anti-PRLR Mouse Monoclonal Antibody [clone: 218616]
Antibodies
Agonist Activity

Type: Primary
Antigen: PRLR
Clonality: Monoclonal
Clone: 218616
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR