Anti-BMP2 Mouse Monoclonal Antibody [clone: 208301]

Supplier: R & D Systems
MAB111
220018-804EA 763.57 USD
220018-804
Anti-BMP2 Mouse Monoclonal Antibody [clone: 208301]
Antibodies
Type: Primary
Antigen: BMP2
Clonality: Monoclonal
Clone: 208301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR