Anti-KLK5 Mouse Monoclonal Antibody [clone: 325302]

Supplier: R & D Systems
MAB11082
220018-802EA 594.37 USD
220018-802
Anti-KLK5 Mouse Monoclonal Antibody [clone: 325302]
Antibodies
Type: Primary
Antigen: KLK5
Clonality: Monoclonal
Clone: 325302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR