Anti-KLK5 Mouse Monoclonal Antibody [clone: 193318]

Supplier: R & D Systems
MAB1108
220018-800EA 772.25 USD
220018-800
Anti-KLK5 Mouse Monoclonal Antibody [clone: 193318]
Antibodies
Type: Primary
Antigen: KLK5
Clonality: Monoclonal
Clone: 193318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR