Anti-SPINT2 Rat Monoclonal Antibody [clone: 170317]

Supplier: R & D Systems
MAB1107
220018-799EA 763.57 USD
220018-799
Anti-SPINT2 Rat Monoclonal Antibody [clone: 170317]
Antibodies
Type: Primary
Antigen: SPINT2
Clonality: Monoclonal
Clone: 170317
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR