Anti-FGL1 Mouse Monoclonal Antibody [clone: 160916]

Supplier: R & D Systems
MAB1105
220018-797EA 763.57 USD
220018-797
Anti-FGL1 Mouse Monoclonal Antibody [clone: 160916]
Antibodies
Type: Primary
Antigen Symbol: FGL1
Antigen Name: fibrinogen-like 1
Antigen Synonyms: Frep1, Lfire1
Clonality: Monoclonal
Clone: 160916
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR