Anti-F11R Mouse Monoclonal Antibody [clone: 654806]

Supplier: R & D Systems
MAB1103
102885-234EA 594.37 USD
102885-234
Anti-F11R Mouse Monoclonal Antibody [clone: 654806]
Antibodies
Type: Primary
Antigen: F11R
Clonality: Monoclonal
Clone: 654806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR