Anti-IL20 Mouse Monoclonal Antibody [clone: 158633]

Supplier: R & D Systems
MAB11022
220018-796EA 772.25 USD
220018-796
Anti-IL20 Mouse Monoclonal Antibody [clone: 158633]
Antibodies
Type: Primary
Antigen: IL20
Clonality: Monoclonal
Clone: 158633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR