Anti-IL20 Mouse Monoclonal Antibody [clone: 158609]

Supplier: R & D Systems
MAB1102
220018-794EA 763.57 USD
220018-794
Anti-IL20 Mouse Monoclonal Antibody [clone: 158609]
Antibodies
Type: Primary
Antigen: IL20
Clonality: Monoclonal
Clone: 158609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR