Anti-ENG Mouse Monoclonal Antibody [clone: 166709]

Supplier: R & D Systems
MAB1097
220018-789EA 763.57 USD
220018-789
Anti-ENG Mouse Monoclonal Antibody [clone: 166709]
Antibodies
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 166709
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR