Anti-EDA2R Mouse Monoclonal Antibody [clone: 173103]

Supplier: R & D Systems
MAB10931
220018-781EA 763.57 USD
220018-781
Anti-EDA2R Mouse Monoclonal Antibody [clone: 173103]
Antibodies
Type: Primary
Antigen: EDA2R
Clonality: Monoclonal
Clone: 173103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR