Anti-SIGIRR Rat Monoclonal Antibody [clone: 161917]

Supplier: R & D Systems
MAB1092
220018-779EA 763.57 USD
220018-779
Anti-SIGIRR Rat Monoclonal Antibody [clone: 161917]
Antibodies
Type: Primary
Antigen: SIGIRR
Clonality: Monoclonal
Clone: 161917
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR