Anti-PPBP Rat Monoclonal Antibody [clone: 159703]

Supplier: R & D Systems
MAB10911
220018-778EA 763.57 USD
220018-778
Anti-PPBP Rat Monoclonal Antibody [clone: 159703]
Antibodies
Type: Primary
Antigen: PPBP
Clonality: Monoclonal
Clone: 159703
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR