Anti-PPBP Rat Monoclonal Antibody [clone: 159742]

Supplier: R & D Systems
MAB1091
220018-777EA 772.25 USD
220018-777
Anti-PPBP Rat Monoclonal Antibody [clone: 159742]
Antibodies
Type: Primary
Antigen: PPBP
Clonality: Monoclonal
Clone: 159742
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR