Anti-IL22RA2 Mouse Monoclonal Antibody [clone: 875504]

Supplier: R & D Systems
MAB10871
102885-232EA 594.37 USD
102885-232
Anti-IL22RA2 Mouse Monoclonal Antibody [clone: 875504]
Antibodies
Type: Primary
Antigen: IL22RA2
Clonality: Monoclonal
Clone: 875504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR