Anti-ARTN Rat Monoclonal Antibody [clone: 185230]

Supplier: R & D Systems
MAB1085
220018-772EA 779.48 USD
220018-772
Anti-ARTN Rat Monoclonal Antibody [clone: 185230]
Antibodies
Type: Primary
Antigen Symbol: ARTN
Antigen Name: artemin
Antigen Synonyms: neublastin
Clonality: Monoclonal
Clone: 185230
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR