Anti-IFNAR2 Rat Monoclonal Antibody [clone: 237526]

Supplier: R & D Systems
MAB1083
220018-771EA 772.25 USD
220018-771
Anti-IFNAR2 Rat Monoclonal Antibody [clone: 237526]
Antibodies
Type: Primary
Antigen: IFNAR2
Clonality: Monoclonal
Clone: 237526
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR