Anti-CTSV Mouse Monoclonal Antibody [clone: 182021]

Supplier: R & D Systems
MAB10801
220018-770EA 577.02 USD
220018-770
Anti-CTSV Mouse Monoclonal Antibody [clone: 182021]
Antibodies
Type: Primary
Antigen: CTSV
Clonality: Monoclonal
Clone: 182021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR