Anti-CTSV Mouse Monoclonal Antibody [clone: 182016]

Supplier: R & D Systems
MAB1080
220018-769EA 772.25 USD
220018-769
Anti-CTSV Mouse Monoclonal Antibody [clone: 182016]
Antibodies
Type: Primary
Antigen: CTSV
Clonality: Monoclonal
Clone: 182016
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR