Anti-IL36A Mouse Monoclonal Antibody [clone: 162122]

Supplier: R & D Systems
MAB1078
220018-767EA 763.57 USD
220018-767
Anti-IL36A Mouse Monoclonal Antibody [clone: 162122]
Antibodies
Type: Primary
Antigen: IL36A
Clonality: Monoclonal
Clone: 162122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR