Anti-F11R Rat Monoclonal Antibody [clone: 160407]

Supplier: R & D Systems
MAB1077
220018-766EA 763.57 USD
220018-766
Anti-F11R Rat Monoclonal Antibody [clone: 160407]
Antibodies
Type: Primary
Antigen: F11R
Clonality: Monoclonal
Clone: 160407
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR