Anti-CER1 Mouse Monoclonal Antibody [clone: 162836]

Supplier: R & D Systems
MAB1075
220018-765EA 772.25 USD
220018-765
Anti-CER1 Mouse Monoclonal Antibody [clone: 162836]
Antibodies
Type: Primary
Antigen: CER1
Clonality: Monoclonal
Clone: 162836
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR