Anti-JAM2 Mouse Monoclonal Antibody [clone: 156624]

Supplier: R & D Systems
MAB10741
220018-764EA 772.25 USD
220018-764
Anti-JAM2 Mouse Monoclonal Antibody [clone: 156624]
Antibodies
Type: Primary
Antigen: JAM2
Clonality: Monoclonal
Clone: 156624
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR