Anti-JAM2 Mouse Monoclonal Antibody [clone: 156623]

Supplier: R & D Systems
MAB1074
220018-763EA 763.57 USD
220018-763
Anti-JAM2 Mouse Monoclonal Antibody [clone: 156623]
Antibodies
Type: Primary
Antigen: JAM2
Clonality: Monoclonal
Clone: 156623
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR