Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 194128]

Supplier: R & D Systems
MAB10721
220018-761EA 779.48 USD
220018-761
Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 194128]
Antibodies
Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 194128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR