Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 194117]

Supplier: R & D Systems
MAB1072
220018-760EA 763.57 USD
220018-760
Anti-SIGLEC5 Mouse Monoclonal Antibody [clone: 194117]
Antibodies
Type: Primary
Antigen: SIGLEC5
Clonality: Monoclonal
Clone: 194117
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR