Anti-TNFA Rat Monoclonal Antibody [clone: 182312]

Supplier: R & D Systems
MAB10702
220018-759EA 772.25 USD
220018-759
Anti-TNFA Rat Monoclonal Antibody [clone: 182312]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 182312
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR