Anti-TNFA Rat Monoclonal Antibody [clone: 182309]

Supplier: R & D Systems
MAB10701
220018-758EA 763.57 USD
220018-758
Anti-TNFA Rat Monoclonal Antibody [clone: 182309]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 182309
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR