Anti-TNFA Rat Monoclonal Antibody [clone: 182303]

Supplier: R & D Systems
MAB1070
220018-757EA 763.57 USD
220018-757
Anti-TNFA Rat Monoclonal Antibody [clone: 182303]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 182303
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Rhesus Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR