Anti-RETN Rat Monoclonal Antibody [clone: 175322]

Supplier: R & D Systems
MAB1069
220018-756EA 772.25 USD
220018-756
Anti-RETN Rat Monoclonal Antibody [clone: 175322]
Antibodies
Type: Primary
Antigen: RETN
Clonality: Monoclonal
Clone: 175322
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR