Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 553517]

Supplier: R & D Systems
MAB10652
102885-230EA 594.37 USD
102885-230
Anti-ADIPOQ Mouse Monoclonal Antibody [clone: 553517]
Antibodies
Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 553517
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR