Anti-F10 Mouse Monoclonal Antibody [clone: 156106]

Supplier: R & D Systems
MAB1063
220018-751EA 763.57 USD
220018-751
Anti-F10 Mouse Monoclonal Antibody [clone: 156106]
Antibodies
Type: Primary
Antigen: F10
Clonality: Monoclonal
Clone: 156106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR