Anti-PDGFRA Rat Monoclonal Antibody [clone: 189208]

Supplier: R & D Systems
MAB1062
220018-750EA 772.25 USD
220018-750
Anti-PDGFRA Rat Monoclonal Antibody [clone: 189208]
Antibodies
Type: Primary
Antigen: PDGFRA
Clonality: Monoclonal
Clone: 189208
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR