Anti-CCL3 Mouse Monoclonal Antibody [clone: 175024]

Supplier: R & D Systems
MAB1061
220018-749EA 763.57 USD
220018-749
Anti-CCL3 Mouse Monoclonal Antibody [clone: 175024]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 175024
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR