Anti-DCN Rat Monoclonal Antibody [clone: 161026]

Supplier: R & D Systems
MAB1060
220018-748EA 763.57 USD
220018-748
Anti-DCN Rat Monoclonal Antibody [clone: 161026]
Antibodies
Type: Primary
Antigen: DCN
Clonality: Monoclonal
Clone: 161026
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR