Anti-KLRD1 Mouse Monoclonal Antibody [clone: 131412]

Supplier: R & D Systems
MAB1058
220018-746EA 763.57 USD
220018-746
Anti-KLRD1 Mouse Monoclonal Antibody [clone: 131412]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: KLRD1
Clonality: Monoclonal
Clone: 131412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR