Anti-NTRK1 Mouse Monoclonal Antibody [clone: 315104]

Supplier: R & D Systems
MAB1056
220018-745EA 594.37 USD
220018-745
Anti-NTRK1 Mouse Monoclonal Antibody [clone: 315104]
Antibodies
Type: Primary
Antigen: NTRK1
Clonality: Monoclonal
Clone: 315104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR