Anti-TNFSF4 Mouse Monoclonal Antibody [clone: 159408]

Supplier: R & D Systems
MAB1054
220018-742EA 772.25 USD
220018-742
Anti-TNFSF4 Mouse Monoclonal Antibody [clone: 159408]
Antibodies
Type: Primary
Antigen: TNFSF4
Clonality: Monoclonal
Clone: 159408
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR