Anti-S100 A12 Mouse Monoclonal Antibody [clone: 161212]

Supplier: R & D Systems
MAB10522
102885-228EA 584.25 USD
102885-228
Anti-S100 A12 Mouse Monoclonal Antibody [clone: 161212]
Antibodies
Type: Primary
Antigen: S100 A12
Clonality: Monoclonal
Clone: 161212
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR