Anti-S100 A12 Mouse Monoclonal Antibody [clone: 161205]

Supplier: R & D Systems
MAB10521
102885-226EA 594.37 USD
102885-226
Anti-S100 A12 Mouse Monoclonal Antibody [clone: 161205]
Antibodies
Type: Primary
Antigen: S100 A12
Clonality: Monoclonal
Clone: 161205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR