Anti-CTSA Mouse Monoclonal Antibody [clone: 179803]

Supplier: R & D Systems
MAB1049
220018-740EA 772.25 USD
220018-740
Anti-CTSA Mouse Monoclonal Antibody [clone: 179803]
Antibodies
Type: Primary
Antigen: CTSA
Clonality: Monoclonal
Clone: 179803
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR