Anti-SPINT1 Mouse Monoclonal Antibody [clone: 169409]

Supplier: R & D Systems
MAB10481
220018-739EA 772.25 USD
220018-739
Anti-SPINT1 Mouse Monoclonal Antibody [clone: 169409]
Antibodies
Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 169409
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR