Anti-SPINT1 Mouse Monoclonal Antibody [clone: 169417]

Supplier: R & D Systems
MAB1048
220018-738EA 772.25 USD
220018-738
Anti-SPINT1 Mouse Monoclonal Antibody [clone: 169417]
Antibodies
Type: Primary
Antigen: SPINT1
Clonality: Monoclonal
Clone: 169417
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR