Anti-CDKN1A Mouse Monoclonal Antibody [clone: 195720]

Supplier: R & D Systems
MAB1047
220018-737EA 572.68 USD
220018-737
Anti-CDKN1A Mouse Monoclonal Antibody [clone: 195720]
Antibodies
Type: Primary
Antigen: CDKN1A
Clonality: Monoclonal
Clone: 195720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR