Anti-APEX1 Mouse Monoclonal Antibody [clone: 2913]

Supplier: R & D Systems
MAB1044
220018-736EA 594.37 USD
220018-736
Anti-APEX1 Mouse Monoclonal Antibody [clone: 2913]
Antibodies
Type: Primary
Antigen: APEX1
Clonality: Monoclonal
Clone: 2913
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR