Anti-PDGFRB Rat Monoclonal Antibody [clone: 182106]

Supplier: R & D Systems
MAB1042
220018-735EA 763.57 USD
220018-735
Anti-PDGFRB Rat Monoclonal Antibody [clone: 182106]
Antibodies
Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: 182106
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR