Anti-TNFRSF13B Rat Monoclonal Antibody [clone: 166010]

Supplier: R & D Systems
MAB1041
220018-734EA 772.25 USD
220018-734
Anti-TNFRSF13B Rat Monoclonal Antibody [clone: 166010]
Antibodies
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 166010
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR